Product Details

SNP ID
rs6990030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37764055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGTCGCAGCCTTAGTTCTGCC[A/C]CTGGTTCTTTTCTTTCTTTTTTTTT
Phenotype
MIM: 611605 MIM: 604436
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ERLIN2 PubMed Links

Gene Details

Gene
ERLIN2
Gene Name
ER lipid raft associated 2
There are no transcripts associated with this gene.

Gene
PROSC
Gene Name
proline synthetase cotranscribed homolog (bacterial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007198.3 Intron NP_009129.1
XM_017013001.1 Intron XP_016868490.1

View Full Product Details