Product Details

SNP ID
rs13084745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:63201381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTGCCTTTACCTATCACATT[A/G]TCTGAAAGGCTGCTGCTTAGGTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNPR PubMed Links

Gene Details

Gene
SYNPR
Gene Name
synaptoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130003.1 Intron NP_001123475.1
NM_144642.4 Intron NP_653243.1
XM_017005731.1 Intron XP_016861220.1
XM_017005732.1 Intron XP_016861221.1

View Full Product Details