Product Details

SNP ID
rs12100931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:32082052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAGCAAGCAACCTACAGTTCTTG[A/C]TCCTCTGAGAATCTATCGCTGCTGT
Phenotype
MIM: 602680
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP5 PubMed Links

Gene Details

Gene
ARHGAP5
Gene Name
Rho GTPase activating protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030055.1 Intron NP_001025226.1
NM_001173.2 Intron NP_001164.2
XM_005267635.3 Intron XP_005267692.1
XM_005267636.3 Intron XP_005267693.1
XM_017021287.1 Intron XP_016876776.1
XM_017021288.1 Intron XP_016876777.1
Gene
ARHGAP5-AS1
Gene Name
ARHGAP5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details