Product Details

SNP ID
rs12574503
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:36512657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGGAAAGATACCTTGCAAACTA[C/G]GAAGTCCTACGCAGATGGAAATAAT
Phenotype
MIM: 179615 MIM: 602355
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAG1 PubMed Links

Gene Details

Gene
RAG1
Gene Name
recombination activating gene 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000448.2 Intron NP_000439.1
XM_005253041.4 Intron XP_005253098.1
XM_011520250.2 Intron XP_011518552.1
Gene
TRAF6
Gene Name
TNF receptor associated factor 6
There are no transcripts associated with this gene.

View Full Product Details