Product Details

SNP ID
rs12274893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5625803 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCTACCAGCGTCTTCACCTGTCT[C/T]AGAACTTGCTCCTCATCATTCTTCC
Phenotype
MIM: 605684 MIM: 608487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM34 PubMed Links

Gene Details

Gene
TRIM34
Gene Name
tripartite motif containing 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003827.1 Intron NP_001003827.1
NM_021616.5 Intron NP_067629.2
NM_130390.2 Intron NP_569074.2
Gene
TRIM5
Gene Name
tripartite motif containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033034.2 Intron NP_149023.2
NM_033092.2 Intron NP_149083.2
NM_033093.2 Intron NP_149084.2
XM_005253183.2 Intron XP_005253240.1
XM_005253184.2 Intron XP_005253241.1
XM_006718358.2 Intron XP_006718421.1
XM_011520426.2 Intron XP_011518728.1
XM_011520427.1 Intron XP_011518729.1
XM_017018460.1 Intron XP_016873949.1
XM_017018461.1 Intron XP_016873950.1
XM_017018462.1 Intron XP_016873951.1
XM_017018463.1 Intron XP_016873952.1
Gene
TRIM6-TRIM34
Gene Name
TRIM6-TRIM34 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003819.3 Intron NP_001003819.1

View Full Product Details