Product Details
- SNP ID
-
rs11905134
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45536416 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATTGATTAGTGTCAGCACAACAAA[A/C]CAACAATTATAGCATTGATTAGTGT
- Phenotype
-
MIM: 609031
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
EPPIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142204954] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPPIN
- Gene Name
- epididymal peptidase inhibitor
There are no transcripts associated with this gene.
- Gene
- EPPIN-WFDC6
- Gene Name
- EPPIN-WFDC6 readthrough
- Gene
- WFDC6
- Gene Name
- WAP four-disulfide core domain 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080827.1 |
1726 |
Intron |
|
|
NP_543017.1 |
View Full Product Details