Product Details

SNP ID
rs6967352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:138710626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGTTTTTAAAAAGATTGAAATC[C/T]TAACCACATTTCAAAAAACGTTTAG
Phenotype
MIM: 605239 MIM: 611700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V0A4 PubMed Links

Gene Details

Gene
ATP6V0A4
Gene Name
ATPase H+ transporting V0 subunit a4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020632.2 Intron NP_065683.2
NM_130840.2 Intron NP_570855.2
NM_130841.2 Intron NP_570856.2
XM_005250393.1 Intron XP_005250450.1
XM_005250394.3 Intron XP_005250451.1
Gene
SVOPL
Gene Name
SVOP like
There are no transcripts associated with this gene.

View Full Product Details