Product Details

SNP ID
rs10744720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6855049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCTGGATCTGTTAGCCCCAGGA[C/T]AGAGGTGAAGGAAGTCCTCTGTGAT
Phenotype
MIM: 607749 MIM: 139130 MIM: 601447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCA3 PubMed Links

Gene Details

Gene
CDCA3
Gene Name
cell division cycle associated 3
There are no transcripts associated with this gene.

Gene
GNB3
Gene Name
G protein subunit beta 3
There are no transcripts associated with this gene.

Gene
USP5
Gene Name
ubiquitin specific peptidase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098536.1 Intron NP_001092006.1
NM_003481.2 Intron NP_003472.2

View Full Product Details