Product Details

SNP ID
rs12393459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119081769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGGGAAGGCCTGGGAGGTAATAC[C/T]TATGGCTGTAACTGCCAATCTTCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA1210 PubMed Links

Gene Details

Gene
KIAA1210
Gene Name
KIAA1210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020721.1 Intron NP_065772.1
XM_006724774.3 Intron XP_006724837.1
XM_017029688.1 Intron XP_016885177.1
XM_017029689.1 Intron XP_016885178.1

View Full Product Details