Product Details

SNP ID
rs12834696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49606789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAAAACATGTGAATTGAGATTT[C/G]TTTGCTACTAAGAAAGTGAGCGGGC
Phenotype
MIM: 300594 MIM: 300720
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GAGE1 PubMed Links

Gene Details

Gene
GAGE1
Gene Name
G antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040663.2 1251 UTR 3 NP_001035753.1
XM_011543896.2 1251 Intron XP_011542198.1
Gene
GAGE2A
Gene Name
G antigen 2A
There are no transcripts associated with this gene.

View Full Product Details