Product Details

SNP ID
rs12134087
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:161708521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACTCAAGTTTACCAGCCCAAATT[C/T]TTTATCATGGCACAAAAGGCCAAGC
Phenotype
MIM: 606891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FCRLA PubMed Links

Gene Details

Gene
FCRLA
Gene Name
Fc receptor like A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184866.1 Intron NP_001171795.1
NM_001184867.1 Intron NP_001171796.1
NM_001184870.1 Intron NP_001171799.1
NM_001184871.1 Intron NP_001171800.1
NM_001184872.1 Intron NP_001171801.1
NM_001184873.1 Intron NP_001171802.1
NM_032738.3 Intron NP_116127.3
XM_006711581.3 Intron XP_006711644.1
XM_011510064.1 Intron XP_011508366.1
XM_011510065.1 Intron XP_011508367.1
XM_011510066.1 Intron XP_011508368.1

View Full Product Details