Product Details

SNP ID
rs13306059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109086608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGTCTCTGTGTGTGAGAGAGC[A/G]AGCGAGCGTGTATGTTCTCATTAGG
Phenotype
MIM: 188545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRHR PubMed Links

Gene Details

Gene
TRHR
Gene Name
thyrotropin releasing hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003301.5 864 Intron NP_003292.1
XM_011517263.2 864 UTR 5 XP_011515565.1

View Full Product Details