Product Details

SNP ID
rs10991378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96498698 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAAATGACCTAAAATATATGAC[A/T]AAGTAGCGAATTGAAAGATATGCCC
Phenotype
MIM: 603505
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDC14B PubMed Links

Gene Details

Gene
CDC14B
Gene Name
cell division cycle 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077181.1 Intron NP_001070649.1
NM_003671.3 Intron NP_003662.1
NM_033331.2 Intron NP_201588.1
XM_005252288.3 Intron XP_005252345.1
XM_011519147.2 Intron XP_011517449.1
XM_011519148.2 Intron XP_011517450.1
XM_011519149.2 Intron XP_011517451.1
XM_011519150.2 Intron XP_011517452.1
XM_011519151.2 Intron XP_011517453.1
XM_011519152.2 Intron XP_011517454.1
XM_011519153.2 Intron XP_011517455.1
XM_011519156.2 Intron XP_011517458.2
XM_011519158.1 Intron XP_011517460.1
XM_011519159.2 Intron XP_011517461.2
XM_017015240.1 Intron XP_016870729.1
XM_017015241.1 Intron XP_016870730.1
XM_017015242.1 Intron XP_016870731.1
XM_017015243.1 Intron XP_016870732.1
XM_017015244.1 Intron XP_016870733.1
XM_017015245.1 Intron XP_016870734.1
XM_017015246.1 Intron XP_016870735.1
XM_017015247.1 Intron XP_016870736.1
XM_017015248.1 Intron XP_016870737.1
XM_017015249.1 Intron XP_016870738.1
Gene
HABP4
Gene Name
hyaluronan binding protein 4
There are no transcripts associated with this gene.

View Full Product Details