Product Details
- SNP ID
-
hCV31101277
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:20189225 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCTTGGGGACCATGAATCACTGG[A/G]AAGTCACCAGAAGGTCTTTAGAAGG
- Phenotype
-
MIM: 616783
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBXN10
PubMed Links
Gene Details
- Gene
- UBXN10
- Gene Name
- UBX domain protein 10
There are no transcripts associated with this gene.
- Gene
- UBXN10-AS1
- Gene Name
- UBXN10 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details