Product Details
- SNP ID
-
rs12033726
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
27
- Location
-
Chr.1:20293639 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTGAGCAAGGCCAAGAAGAGGGTC[A/T]CCTAAACAGGAAGCTGACAGACATC
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
VWA5B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117055222] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- VWA5B1
- Gene Name
- von Willebrand factor A domain containing 5B1
View Full Product Details