Product Details
- SNP ID
-
rs41548215
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31355128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACCTGAACTCTTCCTCCTACACA[T/C]CACAGCAGCGACCACAGCTCCGATG
- Phenotype
-
MIM: 142830
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HLA-B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1051488] are located under a probe and SNP(s) [rs17193040] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HLA-B
- Gene Name
- major histocompatibility complex, class I, B
- Gene
- MIR6891
- Gene Name
- microRNA 6891
There are no transcripts associated with this gene.
View Full Product Details