Product Details

SNP ID
rs41562117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31355152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACAGCAGCGACCACAGCTCCGA[T/C]GACCACAACTGCTAGGACAGCCAGG
Phenotype
MIM: 142830
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
HLA-B PubMed Links

Gene Details

Gene
HLA-B
Gene Name
major histocompatibility complex, class I, B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005514.7 1054 Missense Mutation NP_005505.2
XM_011514557.1 1054 Intron XP_011512859.1
Gene
MIR6891
Gene Name
microRNA 6891
There are no transcripts associated with this gene.

View Full Product Details