Product Details

SNP ID
rs13142404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76224358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAACTTAGAGCCAGGCACCTTA[G/T]AGTTAGGCTCCTGCCCTCCCACAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM47E PubMed Links

Gene Details

Gene
FAM47E
Gene Name
family with sequence similarity 47 member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136570.2 Intron NP_001130042.1
NM_001242936.1 Intron NP_001229865.1

View Full Product Details