Product Details

SNP ID
rs11739799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:110740246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATTCTTCTGGAATTGTTAGAATA[A/T]TAGATTAATGGTATTAATATTAAAT
Phenotype
MIM: 610826
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC25A46 PubMed Links

Gene Details

Gene
SLC25A46
Gene Name
solute carrier family 25 member 46
There are no transcripts associated with this gene.

Gene
TMEM232
Gene Name
transmembrane protein 232
There are no transcripts associated with this gene.

View Full Product Details