Product Details

SNP ID
rs11892277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74420541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTCTTGCAGAACTCAAAATCCCT[G/T]AAGTCTCATTAACCCTATCCACCAG
Phenotype
MIM: 602288
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145054.1 Intron NP_001138526.1
NM_001316764.1 Intron NP_001303693.1
NM_001316765.1 Intron NP_001303694.1
NM_001316766.1 Intron NP_001303695.1
Gene
RTKN
Gene Name
rhotekin
There are no transcripts associated with this gene.

Gene
WDR54
Gene Name
WD repeat domain 54
There are no transcripts associated with this gene.

View Full Product Details