Product Details
- SNP ID
-
hCV31182391
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:32379639 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCACTGTCTTCCCTAAAGTCTTA[A/G]GTTTGATTTGCTCAGTGTAGATATA
- Phenotype
-
MIM: 605638
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BIRC6
PubMed Links
Gene Details
- Gene
- BIRC6
- Gene Name
- baculoviral IAP repeat containing 6
There are no transcripts associated with this gene.
View Full Product Details