Product Details

SNP ID
rs11066984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109044258 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGTTTAACAGGTACAGAATTTTAG[C/T]TTTGCAGATGGAGAGTTCTGGAGAT
Phenotype
MIM: 612492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP30 PubMed Links

Gene Details

Gene
USP30
Gene Name
ubiquitin specific peptidase 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301175.1 Intron NP_001288104.1
NM_032663.4 Intron NP_116052.2
XM_005253962.3 Intron XP_005254019.1
XM_005253965.4 Intron XP_005254022.1
XM_006719653.3 Intron XP_006719716.1
XM_011538894.2 Intron XP_011537196.1
XM_017020048.1 Intron XP_016875537.1
XM_017020049.1 Intron XP_016875538.1
XM_017020050.1 Intron XP_016875539.1
XM_017020051.1 Intron XP_016875540.1
XM_017020052.1 Intron XP_016875541.1
XM_017020053.1 Intron XP_016875542.1
XM_017020054.1 Intron XP_016875543.1
Gene
USP30-AS1
Gene Name
USP30 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details