Product Details

SNP ID
rs13096338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14115204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTATTCACTATAAGGGGCTGTAA[A/C]GCCAAATGCATCATATGGTATACCT
Phenotype
MIM: 611077 MIM: 612048
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHCHD4 PubMed Links

Gene Details

Gene
CHCHD4
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098502.1 Intron NP_001091972.1
NM_144636.2 Intron NP_653237.1
Gene
TMEM43
Gene Name
transmembrane protein 43
There are no transcripts associated with this gene.

View Full Product Details