Product Details

SNP ID
rs10305853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:147482526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGATTCCTGGAAATGAGCTACA[G/T]ACCCACCTTGGAAGAACCTGCAGTT
Phenotype
MIM: 131243
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EDNRA PubMed Links

Gene Details

Gene
EDNRA
Gene Name
endothelin receptor type A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166055.1 Intron NP_001159527.1
NM_001256283.1 Intron NP_001243212.1
NM_001957.3 Intron NP_001948.1

View Full Product Details