Product Details
- SNP ID
-
rs13128636
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70441887 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCACCTATTAATACAATTGAGTG[A/G]TGACCTTTAGTCAATACCCAGTTAT
- Phenotype
-
MIM: 158375
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MUC7
PubMed Links
Gene Details
- Gene
- MUC7
- Gene Name
- mucin 7, secreted
View Full Product Details