Product Details
- SNP ID
-
rs12509294
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:103026309 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCTTAAACATTACATATTTCAATA[C/T]GCATCTTGAAAAAAGTAGCAGCTTT
- Phenotype
-
MIM: 611527
MIM: 611789
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC9B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111508094] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC9B1
- Gene Name
- solute carrier family 9 member B1
There are no transcripts associated with this gene.
- Gene
- SLC9B2
- Gene Name
- solute carrier family 9 member B2
View Full Product Details