Product Details

SNP ID
rs13022122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:158460091 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAGCTAAATATCCAATTATTT[G/T]AATTTTTACCTGTATTGTTTTTGGA
Phenotype
MIM: 604276
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC148 PubMed Links

Gene Details

Gene
CCDC148
Gene Name
coiled-coil domain containing 148
There are no transcripts associated with this gene.

Gene
PKP4
Gene Name
plakophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005476.2 Intron NP_001005476.1
NM_001304969.1 Intron NP_001291898.1
NM_001304970.1 Intron NP_001291899.1
NM_001304971.1 Intron NP_001291900.1
NM_003628.4 Intron NP_003619.2
XM_011512016.1 Intron XP_011510318.1
XM_011512017.1 Intron XP_011510319.1
XM_011512018.1 Intron XP_011510320.1
XM_011512019.1 Intron XP_011510321.1
XM_011512020.2 Intron XP_011510322.1
XM_011512021.2 Intron XP_011510323.1
XM_011512022.1 Intron XP_011510324.1
XM_011512025.1 Intron XP_011510327.1
XM_011512026.2 Intron XP_011510328.1
XM_017005122.1 Intron XP_016860611.1
XM_017005123.1 Intron XP_016860612.1
XM_017005124.1 Intron XP_016860613.1
XM_017005125.1 Intron XP_016860614.1
XM_017005126.1 Intron XP_016860615.1
XM_017005127.1 Intron XP_016860616.1
XM_017005128.1 Intron XP_016860617.1
XM_017005129.1 Intron XP_016860618.1
XM_017005130.1 Intron XP_016860619.1
XM_017005131.1 Intron XP_016860620.1
XM_017005132.1 Intron XP_016860621.1
XM_017005133.1 Intron XP_016860622.1
XM_017005134.1 Intron XP_016860623.1
XM_017005135.1 Intron XP_016860624.1

View Full Product Details