Product Details

SNP ID
rs12026886
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:78649989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTGACAGTGTCACTTGCCAAT[C/G]TCAGTTTTTGTTTTAATTTAAAAAA
Phenotype
MIM: 610468 MIM: 613975
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFI44 PubMed Links

Gene Details

Gene
IFI44
Gene Name
interferon induced protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006417.4 Intron NP_006408.3
XM_005270380.3 Intron XP_005270437.1
XM_011540516.2 Intron XP_011538818.1
XM_017000083.1 Intron XP_016855572.1
Gene
IFI44L
Gene Name
interferon induced protein 44 like
There are no transcripts associated with this gene.

View Full Product Details