Product Details
- SNP ID
-
rs11162434
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:78233910 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTTAGGATGTTGGCAATTACCCCA[C/G]AAATTGTCTTTGAAAAAAGTTGAAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MGC27382
PubMed Links
Gene Details
- Gene
- MGC27382
- Gene Name
- uncharacterized MGC27382
There are no transcripts associated with this gene.
View Full Product Details