Product Details

SNP ID
rs10889296
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:62198196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAACCAGGCACTTTATCTTGGT[A/G]TGGTAGCATGCCTGTAATACCAGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
L1TD1 PubMed Links

Gene Details

Gene
L1TD1
Gene Name
LINE1 type transposase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details