Product Details

SNP ID
rs12126777
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:43281875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGAAAAATAAAAATGAGTGACA[C/G]CTTTTTATTTTTATTCAAAATGAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1orf210 PubMed Links

Gene Details

Gene
C1orf210
Gene Name
chromosome 1 open reading frame 210
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164829.1 1545 Intron NP_001158301.1
NM_182517.2 1545 Intron NP_872323.1
XM_011540802.1 1545 UTR 3 XP_011539104.1
Gene
TMEM125
Gene Name
transmembrane protein 125
There are no transcripts associated with this gene.

View Full Product Details