Product Details

SNP ID
rs11134516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:168486819 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCGGTGGGCTGGAGCCTGGCAT[A/G]TTCAGAACAGTTCTCTCCCTACAAT
Phenotype
MIM: 107820
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RARS PubMed Links

Gene Details

Gene
RARS
Gene Name
arginyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002887.3 Intron NP_002878.2

View Full Product Details