Product Details

SNP ID
rs11032365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:33746162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAGCCCCAGATAACTTCAGTGAT[C/G]AATCATATCAAACATTTAGGGAAAA
Phenotype
MIM: 107271 MIM: 609089
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD59 PubMed Links

Gene Details

Gene
CD59
Gene Name
CD59 molecule
There are no transcripts associated with this gene.

Gene
FBXO3
Gene Name
F-box protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012175.3 Intron NP_036307.2
NM_033406.2 Intron NP_208385.1
XM_011519980.1 Intron XP_011518282.1
XM_011519981.2 Intron XP_011518283.1
XM_011519982.1 Intron XP_011518284.1
XM_017017533.1 Intron XP_016873022.1

View Full Product Details