Product Details

SNP ID
rs11195939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:112382346 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCAGCAGACTCTCCTCGCTAGTT[A/G]CCTTTCTCTCAAATTGTGCCAATGG
Phenotype
MIM: 605677
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSL5 PubMed Links

Gene Details

Gene
ACSL5
Gene Name
acyl-CoA synthetase long-chain family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016234.3 Intron NP_057318.2
NM_203379.1 Intron NP_976313.1
NM_203380.1 Intron NP_976314.1
XM_017016324.1 Intron XP_016871813.1

View Full Product Details