Product Details

SNP ID
rs6584588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104139413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCTCACGCACACATGTGCATAC[C/T]GCCCCCTATCCCCACCTAGACACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CFAP43 PubMed Links

Gene Details

Gene
CFAP43
Gene Name
cilia and flagella associated protein 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025145.5 Intron NP_079421.5
XM_005270171.2 Intron XP_005270228.1
XM_005270172.3 Intron XP_005270229.1
XM_011540196.2 Intron XP_011538498.1
XM_011540197.2 Intron XP_011538499.1
XM_011540198.2 Intron XP_011538500.1
XM_011540199.2 Intron XP_011538501.1
XM_011540200.2 Intron XP_011538502.1
XM_011540201.2 Intron XP_011538503.1
XM_011540202.2 Intron XP_011538504.1
XM_011540203.2 Intron XP_011538505.1
XM_017016681.1 Intron XP_016872170.1
XM_017016682.1 Intron XP_016872171.1
XM_017016683.1 Intron XP_016872172.1
XM_017016684.1 Intron XP_016872173.1

View Full Product Details