Product Details

SNP ID
rs10786391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:98136167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCCACTGTGCCCGACCTCTGC[A/G]ATTAATTTTTAAAGGGGTAAGAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
R3HCC1L PubMed Links

Gene Details

Gene
R3HCC1L
Gene Name
R3H domain and coiled-coil containing 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256619.1 Intron NP_001243548.1
NM_001256620.1 Intron NP_001243549.1
NM_001256621.1 Intron NP_001243550.1
NM_014472.4 Intron NP_055287.4
NM_138469.2 Intron NP_612478.2
XM_011539639.1 Intron XP_011537941.1
XM_011539640.1 Intron XP_011537942.1
XM_011539641.1 Intron XP_011537943.1
XM_011539642.1 Intron XP_011537944.1
XM_011539643.1 Intron XP_011537945.1
XM_011539644.1 Intron XP_011537946.1
XM_011539645.2 Intron XP_011537947.1
XM_011539646.1 Intron XP_011537948.1
XM_011539647.1 Intron XP_011537949.1
XM_011539648.1 Intron XP_011537950.1
XM_011539649.1 Intron XP_011537951.1
XM_017016074.1 Intron XP_016871563.1
XM_017016075.1 Intron XP_016871564.1
XM_017016076.1 Intron XP_016871565.1
XM_017016077.1 Intron XP_016871566.1
XM_017016078.1 Intron XP_016871567.1
XM_017016079.1 Intron XP_016871568.1
XM_017016080.1 Intron XP_016871569.1
XM_017016081.1 Intron XP_016871570.1
XM_017016082.1 Intron XP_016871571.1
XM_017016083.1 Intron XP_016871572.1

View Full Product Details