Product Details

SNP ID
rs12086086
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:185127444 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTGCAATTCTTGTCACAATGTT[C/G]TTAGGAATCTTGAATTTCTGTAAGA
Phenotype
MIM: 611673
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRMT1L PubMed Links

Gene Details

Gene
TRMT1L
Gene Name
tRNA methyltransferase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202423.1 Intron NP_001189352.1
NM_030934.4 Intron NP_112196.3
XM_011510028.2 Intron XP_011508330.1
XM_017002419.1 Intron XP_016857908.1

View Full Product Details