Product Details

SNP ID
rs12030331
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:200032419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGTGGTTGATAAGTAAGAAAA[A/G]ATATTTGGAGTGTAACTTTTAAACA
Phenotype
MIM: 604453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR5A2 PubMed Links

Gene Details

Gene
NR5A2
Gene Name
nuclear receptor subfamily 5 group A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276464.1 Intron NP_001263393.1
NM_003822.4 Intron NP_003813.1
NM_205860.2 Intron NP_995582.1
XM_005245062.3 Intron XP_005245119.1
XM_011509380.1 Intron XP_011507682.1
XM_011509381.2 Intron XP_011507683.1
XM_011509382.1 Intron XP_011507684.1
XM_011509384.2 Intron XP_011507686.1
XM_017000904.1 Intron XP_016856393.1

View Full Product Details