Product Details
- SNP ID
-
rs12024960
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
29
- Location
-
Chr.1:200553493 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGTACACCTTTATTCTTACTGCC[A/G]TCAATCCCGCTTGATTTAGATTGTT
- Phenotype
-
MIM: 611279
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIF14
PubMed Links
Gene Details
- Gene
- KIF14
- Gene Name
- kinesin family member 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001305792.1 |
4593 |
Silent Mutation |
GAC,GAT |
D1123D |
NP_001292721.1 |
NM_014875.2 |
4593 |
Silent Mutation |
GAC,GAT |
D1614D |
NP_055690.1 |
XM_011510231.2 |
4593 |
Silent Mutation |
GAC,GAT |
D1614D |
XP_011508533.1 |
XM_011510232.2 |
4593 |
Silent Mutation |
GAC,GAT |
D1614D |
XP_011508534.1 |
XM_011510233.2 |
4593 |
Silent Mutation |
GAC,GAT |
D1586D |
XP_011508535.1 |
XM_011510235.2 |
4593 |
Silent Mutation |
GAC,GAT |
D1490D |
XP_011508537.1 |
XM_011510236.1 |
4593 |
Silent Mutation |
GAC,GAT |
D1123D |
XP_011508538.1 |
XM_017003005.1 |
4593 |
Silent Mutation |
GAC,GAT |
D1614D |
XP_016858494.1 |
XM_017003006.1 |
4593 |
Silent Mutation |
GAC,GAT |
D1571D |
XP_016858495.1 |
XM_017003007.1 |
4593 |
Silent Mutation |
GAC,GAT |
D1425D |
XP_016858496.1 |
View Full Product Details