Product Details
- SNP ID
-
rs6957529
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:128965406 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATATCACTTCACACTTGGTGTGAAA[C/T]GCTAAGATGGCTACTCTCAAAAACC
- Phenotype
-
MIM: 610032
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TNPO3
PubMed Links
Gene Details
- Gene
- TNPO3
- Gene Name
- transportin 3
View Full Product Details