Product Details

SNP ID
rs12279926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57674923 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTCTCTCAAGGATCTAGAGACAGT[A/G]CTGCTTTTCTCTGACTTAATATTTT
Phenotype
MIM: 614586
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC5 PubMed Links

Gene Details

Gene
ZDHHC5
Gene Name
zinc finger DHHC-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015457.2 Intron NP_056272.2
XM_011544899.1 Intron XP_011543201.1
XM_011544900.1 Intron XP_011543202.1
XM_011544901.1 Intron XP_011543203.1
XM_017017498.1 Intron XP_016872987.1

View Full Product Details