Product Details

SNP ID
rs11233495
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:83159435 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACAGCGCCTAATATTTAGCAGCT[C/T]AGGTTGTCACTCAAAGAACCTTGCT
Phenotype
MIM: 608876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCF11 PubMed Links

Gene Details

Gene
PCF11
Gene Name
PCF11 cleavage and polyadenylation factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015885.3 Intron NP_056969.2
XM_005274048.1 Intron XP_005274105.1
XM_005274049.1 Intron XP_005274106.1
XM_017017918.1 Intron XP_016873407.1

View Full Product Details