Product Details

SNP ID
rs13038771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:47210635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCCCCGCGCCGGGGGCTCAGG[C/G]TCAACTGAGGGTTTTGTCATTTTCA
Phenotype
MIM: 615713
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZMYND8 PubMed Links

Gene Details

Gene
ZMYND8
Gene Name
zinc finger MYND-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281769.1 4298 Intron NP_001268698.1
NM_001281771.2 4298 UTR 3 NP_001268700.1
NM_001281772.2 4298 UTR 3 NP_001268701.1
NM_001281773.2 4298 UTR 3 NP_001268702.1
NM_001281774.2 4298 UTR 3 NP_001268703.1
NM_001281775.2 4298 UTR 3 NP_001268704.1
NM_001281776.2 4298 UTR 3 NP_001268705.1
NM_001281777.2 4298 UTR 3 NP_001268706.1
NM_001281778.2 4298 UTR 3 NP_001268707.1
NM_001281779.2 4298 UTR 3 NP_001268708.1
NM_001281780.2 4298 UTR 3 NP_001268709.1
NM_001281781.2 4298 UTR 3 NP_001268710.1
NM_001281782.2 4298 UTR 3 NP_001268711.1
NM_001281783.2 4298 UTR 3 NP_001268712.1
NM_001281784.2 4298 UTR 3 NP_001268713.1
NM_012408.5 4298 UTR 3 NP_036540.3
NM_183047.3 4298 UTR 3 NP_898868.1
NM_183048.3 4298 UTR 3 NP_898869.1
XM_005260356.4 4298 UTR 3 XP_005260413.1
XM_005260358.2 4298 UTR 3 XP_005260415.1
XM_005260360.4 4298 UTR 3 XP_005260417.1
XM_005260362.2 4298 UTR 3 XP_005260419.1
XM_005260364.2 4298 UTR 3 XP_005260421.1
XM_005260366.2 4298 UTR 3 XP_005260423.1
XM_006723760.3 4298 UTR 3 XP_006723823.1
XM_006723762.3 4298 UTR 3 XP_006723825.1
XM_006723763.1 4298 UTR 3 XP_006723826.1
XM_006723766.1 4298 UTR 3 XP_006723829.1
XM_011528749.2 4298 UTR 3 XP_011527051.1
XM_011528751.1 4298 UTR 3 XP_011527053.1
XM_011528752.1 4298 UTR 3 XP_011527054.1
XM_011528753.1 4298 UTR 3 XP_011527055.1
XM_011528754.1 4298 UTR 3 XP_011527056.1
XM_017027760.1 4298 UTR 3 XP_016883249.1
XM_017027761.1 4298 UTR 3 XP_016883250.1
XM_017027762.1 4298 UTR 3 XP_016883251.1
XM_017027763.1 4298 UTR 3 XP_016883252.1
XM_017027764.1 4298 UTR 3 XP_016883253.1
XM_017027765.1 4298 UTR 3 XP_016883254.1
XM_017027766.1 4298 UTR 3 XP_016883255.1
XM_017027767.1 4298 UTR 3 XP_016883256.1
XM_017027768.1 4298 UTR 3 XP_016883257.1
XM_017027769.1 4298 UTR 3 XP_016883258.1
XM_017027770.1 4298 UTR 3 XP_016883259.1
XM_017027771.1 4298 UTR 3 XP_016883260.1
XM_017027772.1 4298 UTR 3 XP_016883261.1
XM_017027773.1 4298 UTR 3 XP_016883262.1
XM_017027774.1 4298 UTR 3 XP_016883263.1
XM_017027775.1 4298 UTR 3 XP_016883264.1
XM_017027776.1 4298 UTR 3 XP_016883265.1
XM_017027777.1 4298 UTR 3 XP_016883266.1
XM_017027778.1 4298 Intron XP_016883267.1
XM_017027779.1 4298 UTR 3 XP_016883268.1
XM_017027780.1 4298 UTR 3 XP_016883269.1

View Full Product Details