Product Details

SNP ID
rs12884398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50107240 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCACTGGTCCTAACCATCAGTGG[C/T]CGAATCTTAGAATGTGAGGATTGGC
Phenotype
MIM: 601247 MIM: 615260
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01599 PubMed Links

Gene Details

Gene
LINC01599
Gene Name
long intergenic non-protein coding RNA 1599
There are no transcripts associated with this gene.

Gene
SOS2
Gene Name
SOS Ras/Rho guanine nucleotide exchange factor 2
There are no transcripts associated with this gene.

Gene
VCPKMT
Gene Name
valosin containing protein lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040662.1 Intron NP_001035752.1
NM_024558.2 Intron NP_078834.2
XM_005268060.3 Intron XP_005268117.1
XM_017021640.1 Intron XP_016877129.1
XM_017021641.1 Intron XP_016877130.1
XM_017021642.1 Intron XP_016877131.1

View Full Product Details