Product Details

SNP ID
rs11622124
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:36678762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTATAATTTTTTTCTGGTTCTTG[G/T]ATTTTAAAAAATCTAATATTAATGG
Phenotype
MIM: 167416 MIM: 607571
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PAX9 PubMed Links

Gene Details

Gene
PAX9
Gene Name
paired box 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006194.3 3106 Intron NP_006185.1
Gene
SLC25A21
Gene Name
solute carrier family 25 member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171170.1 3106 Intron NP_001164641.1
NM_030631.3 3106 UTR 3 NP_085134.1
XM_011537287.2 3106 Intron XP_011535589.1
XM_011537288.2 3106 Intron XP_011535590.1
XM_011537289.2 3106 Intron XP_011535591.1

View Full Product Details