Product Details
- SNP ID
-
rs12200397
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:36496462 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCAGATTACCTAACACTTAGTAA[A/G]TAACAAAAGAACAGATTTTTGTGAC
- Phenotype
-
MIM: 615932
MIM: 606964
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCTD20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77908513] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCTD20
- Gene Name
- potassium channel tetramerization domain containing 20
There are no transcripts associated with this gene.
- Gene
- STK38
- Gene Name
- serine/threonine kinase 38
View Full Product Details