Product Details

SNP ID
rs4082950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48210189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCAAGACATGGAAAGGGCAGAAA[C/G]AAATTTAGGGCATTATAAAAAGATA
Phenotype
MIM: 600839
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CTXN2 PubMed Links

Gene Details

Gene
CTXN2
Gene Name
cortexin 2
There are no transcripts associated with this gene.

Gene
SLC12A1
Gene Name
solute carrier family 12 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000338.2 Intron NP_000329.2
NM_001184832.1 Intron NP_001171761.1
XM_005254606.1 Intron XP_005254663.1

View Full Product Details