Product Details

SNP ID
rs12357032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:79075297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCAGATGGGGAAACTTAGGCCC[A/G]CAGAGGGACAGAGACTGGCTCAGGA
Phenotype
MIM: 607159
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZMIZ1 PubMed Links

Gene Details

Gene
ZMIZ1
Gene Name
zinc finger MIZ-type containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020338.3 Intron NP_065071.1
XM_005269987.4 Intron XP_005270044.1
XM_005269988.3 Intron XP_005270045.1
XM_006717923.3 Intron XP_006717986.1
XM_006717924.3 Intron XP_006717987.1
XM_006717925.3 Intron XP_006717988.1
XM_011539978.2 Intron XP_011538280.1
XM_011539979.1 Intron XP_011538281.1
XM_011539980.2 Intron XP_011538282.1
XM_017016440.1 Intron XP_016871929.1
XM_017016441.1 Intron XP_016871930.1
XM_017016442.1 Intron XP_016871931.1
XM_017016443.1 Intron XP_016871932.1
Gene
ZMIZ1-AS1
Gene Name
ZMIZ1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details