Product Details

SNP ID
rs12378820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136007823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGGGGAGCCCAGGCAGTACACG[C/G]CTGGGTGGGGTGGGGGGACTGGGCC
Phenotype
MIM: 615786
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NACC2 PubMed Links

Gene Details

Gene
NACC2
Gene Name
NACC family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144653.4 5613 UTR 3 NP_653254.1
XM_011518221.2 5613 UTR 3 XP_011516523.1

View Full Product Details