Product Details

SNP ID
rs11505819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95584467 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCATTTCAGGTGTCATTCATTA[G/T]CTCTCAGTCACAACAACTGACAAAT
Phenotype
MIM: 602527
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDK4 PubMed Links

Gene Details

Gene
PDK4
Gene Name
pyruvate dehydrogenase kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002612.3 2730 UTR 3 NP_002603.1

View Full Product Details